Elbow connecting tube, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1175-15 R1175-50
BOHLR1175-15EA 106 EUR
BOHLR1175-15 BOHLR1175-50
Elbow connecting tube, b.safe
Fittingen en connectoren
Elbow tube approx. 90° made of static dissipative PE-EX.
  • Available in two different lengths
With lock nut and clamp ring for inserting and clamping b.safe Waste pipes with Ext.Ø 32 mm, end piece with Ext.Ø 32 mm.
Order Now
Specificaties
Environmentally Preferable