T-connection tubes for waste pipes, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1194-01 R1194-02
BOHLR1194-01EA 261 EUR
BOHLR1194-01 BOHLR1194-02
T-connection tubes for waste pipes, b.safe
Fittingen en connectoren
T-shaped connector made of static dissipative PE-EX.
Lateral connections with union nut and clamp ring to insert and clamp b.safe waste pipes with Ext.Ø 32 mm. Central connection with male thread GL 45 to mount e.g. b.safe system caps.
Order Now
Specificaties
Environmentally Preferable