Cryogenic Multi-Colour Dots for Tops of 1,5 ml Microtubes (Writable)
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JTA-13X6A JTA-2610A JTA-2610BE JTA-2610BL JTA-2610BR JTA-2610GA JTA-2610GS JTA-2610LA JTA-2610LE JTA-2610MT JTA-2610OR JTA-2610PI JTA-2610RE JTA-2610SI JTA-2610VI JTA-2610WH JTA-2610YE
GAINJTA-13X6AEA 20.2 EUR
GAINJTA-13X6A GAINJTA-2610A GAINJTA-2610BE GAINJTA-2610BL GAINJTA-2610BR GAINJTA-2610GA GAINJTA-2610GS GAINJTA-2610LA GAINJTA-2610LE GAINJTA-2610MT GAINJTA-2610OR GAINJTA-2610PI GAINJTA-2610RE GAINJTA-2610SI GAINJTA-2610VI GAINJTA-2610WH GAINJTA-2610YE
Cryogenic Multi-Colour Dots for Tops of 1,5 ml Microtubes (Writable)
Etiketten
Writable cryo colour dots and rectangles for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen Dewars. Easy to write on with permanent cryo markers, these labels are available in a variety of colours.
  • Temperature range: −196 to +110 °C
  • Available as a multi-color sheet for colour coding and easy identification
  • Small round labels are ideal for 0,5 to 1,5 ml Eppendorf tube caps and vials
  • Strong adhesive ensures labels won’t wrinkle or peel during long-term storage in −80 °C freezers, dry ice, and liquid nitrogen at −196 °C
Ideal for identification of vials, tops, cans, plastic bags, and other containers for long-term storage and colour-coding in cryogenic and harsh laboratory environments.