Mini-display table level sensor, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R659-01 R659-02 R659-03
BOHLR659-01EA 221 EUR
BOHLR659-01 BOHLR659-02 BOHLR659-03
Mini-display table level sensor, b.safe
Niveausensoren
The b.safe Mini-display is the optimal basic solution for the local control of single HPLC units.
  • With plugs for EU, Australia, USA, Japan, and UK
  • Compatible with any version of b.safe waste caps
  • Additional comfort provides the possibility to snooze the alarm tone during the canister exchange
Display unit is designed to monitor connected fill-level sensors in b.safe Waste Caps and b.safe System Waste Caps. Tight plastic casing allows secure placement in the work area. Easy operation via touch surface.
Alarm box in plastic housing. Front side withMore Product Information