Waste tubes, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1410-06 R1410-10 R1410-15 R1410-25
BOHLR1410-06EA 117 EUR
BOHLR1410-06 BOHLR1410-10 BOHLR1410-15 BOHLR1410-25
Waste tubes, b.safe
Slangen Transfer Tubing
Flexible corrugated tubing to lead liquid solvent waste e.g. from a b.safe Desktop Mount into a canister with Waste Cap and connection GL 25.
  • Corrugated tubes in static dissipative PTFE-EX
  • Highly flexible
  • High chemical resistance
Cut from flexible corrugated tubing made of static dissipative PTFE-EX, spiral-shaped inside and outside. Including two b.safe Screw Joints for Waste Tube fitting connection thread GL 25 of e.g. b.safe Desktop Mounts or b.safe Waste Caps, 1 hexagon key SW10 and 1 tubing holder for wall mounting. To be able to cut the tubingMore Product Information
Order Now
Specificaties
Environmentally Preferable