Waste tubes, smooth, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1420-06 R1420-10 R1420-15 R1420-25
BOHLR1420-06EA 167 EUR
BOHLR1420-06 BOHLR1420-10 BOHLR1420-15 BOHLR1420-25
Waste tubes, smooth, b.safe
Slangen Transfer Tubing
Flexible corrugated tubing to lead liquid solvent waste e.g. from a b.safe Desktop Mount into a canister with b.safe Waste Cap and connection GL 25.
  • Corrugated tubes in static dissipative PTFE-EX
  • Highly flexible
  • High chemical resistance
Cut from flexible corrugated tubing made of static dissipative PTFE-EX, spiral-shaped outside and smooth inside. Including two b.safe Screw Joints for Waste Tube Smooth fitting connection thread GL 25 of e.g. b.safe Desktop Mounts or b.safe Waste Caps and 1 tubing holder for wall mounting. To be able to cut the tubing to the required length, the b.safe Screw Joints are not mounted.
If longer tubing is required, b.safe Corrugated Tubing R 773-16 and b.safe Screw Joints for Waste Tube R 1405-06 can be combined individually.