Vacuum filtration units, Sartolab® BT and RF
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
180E12-E 180F02-E 180D03-E 180E03-E 180E13-E 180F03-E 180E04-E 180E14-E 180F04-E 180D05-E 180E05-E 180E15-E 180F15-E 180F05-E 180E02-E
514-1307EA 55 EUR
514-1307 514-1316 514-1302 514-1304 514-1308 514-1317 514-1305 514-1309 514-1318 514-1303 514-1306 514-1310 514-1330 514-1319 514-1341
Vacuum filtration units, Sartolab® BT and RF
Filters Flesfilters
These bottle-top filters and vacuum filtration units are single-use units for the sterile filtration of samples.
  • Highest flow rates with a large filtration area
  • No loss of protein with a low protein affinity membrane
  • Low hold-up volume due to an optimised membrane support
  • Can be used as stand-alone system or with the Sartolab® Multistation, which allows parallel filtration of up to six samples
Sartolab® BT bottle-top filters are comprised of:
A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
A tube connector for vacuum connection (for stand-alone filtration)
Sartolab® RF vacuum filtration unit is comprised of:
A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
A bottle, with graduation and writing field
A tube connector for vacuum connection (for stand-alone filtration)
A screw cap for storage of the filtrate (individually wrapped to maintain sterility) history