Control Waste Cap DLC, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M250-30 M255-30 M256-30
BOHLM250-30EA 546 EUR
BOHLM250-30 BOHLM255-30 BOHLM256-30
Control Waste Cap DLC, b.safe
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
To avoid spill over of canisters through the connection of a b.safe Control Waste Cap to a b.safe Display for electronic level control.
  • PPS screw cap
  • Freely rotatable insert made of PTFE
  • Avoid overflowing canisters
With 2 permanently fixed cables (length approx. 0,5 m each) for the connection to a b.safe Display which signals a pre-alert when approx. 50% of the nominal volume are reached and a main alert for canister change as soon as 100% of the nominal volume are reached. With 1 additional red LED flashing light for easy identification of a fullMore Product Information