Grade 0858 ½ qualitative filter papers, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10334345 10334347 10334351 10334352 10334353
512-0520EA 39.6 EUR
512-0520 512-0521 512-0522 512-0523 512-0409
Grade 0858 ½ qualitative filter papers, Whatman™
Filters Filterpapier Kwalitatief filterpapier
Grained, with medium fast flow rate and medium retention. A universal filter paper, used for the filtration of extracts, oils, beer, syrups etc. Also applied in filter presses or for the aspiration of liquids.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
More Product Information
Order Now
Specificaties
Nr 0858 ½
Environmentally Preferable
Gewicht 75 g/m²
Dikte 170 µm
Partikelretentie 7 - 12 µm
Filtratiesnelheid 55 s (Herzberg)