Safety spectacles, Seez™
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1SEZBG23CKN 1SEZBG23SKN 1SEZLGB23CKN
JSPA1SEZBG23CKNEA 19.4 EUR
JSPA1SEZBG23CKN JSPA1SEZBG23SKN JSPA1SEZLGB23CKN
Safety spectacles, Seez™
Brillen
Wraparound style safety spectacle with integrated browguard for comfort and protection.
  • Large field of vision
  • Moulded browguard
  • Flexible nose bridge
  • Polycarbonate lens
  • Flexofit temple with memory
  • Fully adjustable
  • Soft TPR temple tips
This exceptional eyewear concept unites innovative technologies and high-tech features and benefits. Wraparound style, 10 lens base curve provides amazing optical clarity, along with an integrated browguard for comfort and protection.
Swiss One Flexofit technology, non-slip side arms utilise shape memory material which can be formed to provide optimal fit and security. A flexible nose bridge provides added comfort and grip.
Certificaten: Meets EN 166:2001.