Splash guard, without impact protection, Force®8
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JSPAAFN060-000-000EA 26.8 EUR
JSPAAFN060-000-000
Splash guard, without impact protection, Force®8
Gelaatsschermen
The Force®8 Splash Guard can be fitted to a Force®8 half mask to reduce the likelihood of exhaled bacteria/virus hitting the wearer's face.
  • Easily fits to JSP Force®8 half mask
  • Provides a layer of protection to block aerosols and droplets
  • Prevents wearer from touching eyes and face
  • Quick and simple cleaning with soap and water
The Force®8 Splash Guard prevents the wearer from touching eyes and face. Quick and simple cleaning can be performed with soap and water.