Gas goggle, non-vented, EVO®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EVO™
111-2596EA 24 EUR
111-2596
Gas goggle, non-vented, EVO®
Ruimzichtbrillen
Single lens goggle for everyday protection.
  • PVC frame
  • Elastic headband
  • Anti-mist
  • Molten metal protection
  • Non vented design
  • High level of protection
Standard single lens goggle for everyday protection, engineered with anti-mist and scratch resistant single lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Goggles offer dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Certificaten: EN 166; EN 170.