Horizontal fold flat dust masks, 0300 Series, JSP LTD
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JSPABGT170-000-S00EA 0 EUR
JSPABGT170-000-S00
Horizontal fold flat dust masks, 0300 Series, JSP LTD
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
Fold flat horizontal disposable mask complete with an adjustable nose clip and 3600 comfort foam seal. Valved and unvalved options are available.
  • Fold flat design
  • Adjustable nose clip
  • Foam padded nose bridge
  • Ratchet adjustment for an effective facial fit
The fold flat design is compact and easy to carry in a pocket. Ratchet adjustment on nape strap helps to achieve a secure facial fit. Masks also feature a mouldable nose bridge and foam pad on the inside for comfort and a firmer fit. Adjustable nose clip aids in achieving a comfortable face seal. Low resistance exhalation valve allows for easy breathing.
Minimum filtration efficiency: 99% for FFP3 rated, 94% for FFP2 rated.
Certificaten: EN149.