Vertical fold flat disposable masks, F600 series
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BGV120-000-Q00 BGW170-000-S00
111-2582EA 24.1 EUR
111-2582 111-2583
Vertical fold flat disposable masks, F600 series
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
Vertical fold flat respirator with foam padded adjustable nose clip and extended chin coverage.
  • Folds flat
  • Foam padded nose bridge
  • Toggle adjustment
  • Individually packaged
A moldable nose bridge and foam pad on the inside provide comfort and a firmer fit. Metal-free toggle adjuster on each strap ensures an effective facial fit. Fold flat design is compact and easy to carry in a pocket. Packaged individually in a clear resealable bag. Valved and unvalved options are available.
Minimum filtration efficiency: 99% for FFP3 rated, 94% for FFP2 rated.
Certificaten: EN 149:2001 + A1:2009.