Half masks with Typhoon™ valve, Force®8
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BHG003-1L5-000 BHG003-2L5-000 BHT003-0L5-000 BHG002-0L5-000
JSPABHG003-1L5-000EA 40.5 EUR
JSPABHG003-1L5-000 JSPABHG003-2L5-000 JSPABHT003-0L5-000 JSPABHG002-0L5-000
Half masks with Typhoon™ valve, Force®8
Ademhalingsmaskers Herbruikbare maskers Halfgelaatsmaskers
The Force®8 twin cartridge half mask with Typhoon™ valve offers superior low breathing resistance and a 4-point suspension harness with quick release buckles.
  • Durable thermoplastic rubber mask
  • Offers a superior fit to most face shapes
  • CR2 reflectivity
  • 4-point cradle suspension harness
  • Mask accepts the full range of low profile Force®8 filters
The Force®8 twin cartridge half mask with Typhoon™ valve is made with a durable thermoplastic rubber for a superior fit to most face shapes. It accepts the full range of low profile Force®8 filters, giving theMore Product Information