PAPR systems, PowerCap® Infinity®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CAU660-000-400 CEA646-000-100
JSPACAU660-000-400EA 94.6 EUR
JSPACAU660-000-400 JSPACEA646-000-100
PAPR systems, PowerCap® Infinity®
Ademhalingsmaskers Ademhalingssystemen
A fully integrated TH3 powered air respirator offering four of the highest levels of protection in one intuitive head-mounted unit. The system provides complete above the neck 4-in-1 protection; respiratory, head, eye, and face.
  • 4-in-1 protection
  • Super strong ABS shell
  • Superior all-day protection
  • Optical class 1 visor
  • High capacity HEPA filters
  • Polycarbonate face shield
  • Rubber skirt
PowerCap® Infinity® is the perfect choice for many industries where dust poses a great danger to health. PowerCap® Infinity® removes the need for face fit testing, ensuring that a high volume of clean air is delivered to the breathing zone. High capacity HEPA filters coupled with aerodynamically optimised pre-filter covers ensure PowerCap® Infinity® delivers 160 litres per minute of clean air to the user with maximum battery efficiency. Filters can be changed easily with the quick-change bayonet system.
The optical class 1 visor offers impact protection at speeds of up to 270 mph. The visor has a replaceable peel-off visor protector to prolong its life. Developed to seamlessly integrate with Sonis® helmet-mounted ear defenders.
Certificaten: Respiratory protection: EN12941:1998 + A2:2008 TH3P, APF 40. Head protection: EN397. Eye and face protection: EN166.1.B. Hearing protection (Optional extra): EN352-3. Approval: AS/NZS 1716:2012, AS/NZS 1337.1, AS/NZS 1801.