Barrier with anti-trip feet, 2 m, Titan®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Titan™
JSPAKBP073-300-200EA 197 EUR
JSPAKBP073-300-200
Barrier with anti-trip feet, 2 m, Titan®
Verkeersveiligheid Verkeerskegels/-hekken
Easy to use, assemble, and store barrier system available in 2 metre lengths.
  • Made from the strongest virgin materials available
  • Added vertical bars for increased rigidity
  • 100% metal-free and fully recyclable
  • Fully reflective panel meeting EN 12899-1 requirements
The Titan® is the recognised and well-established stacking barrier from JSP with many unique features. It has been modified to a one-piece injection moulded design made from prime grade virgin polypropylene, offering sub-zero temperature and 2-year colour fastness properties. The new design is 100% metal-free and fully recyclable, with a fully reflective panel meeting EN 12899-1 requirements for higher visibility in poor light conditions.
The clever design allows for simple and practical storing, stacking, and unstacking. Water apertures have been added to the one-piece mould so that when they are stacked they do not collect water within the frames and therefore do not add extra weight or inconvenience. Flexi-coupling clips enable easy connecting and compatibility with not only other styles of JSP barriers but also other brands. The clip can be conveniently retained to provide added rigidity.
The Anti-trip feet with a new scalloped design prevent pedestrian/worker accidents, and feature two reflectors per foot for extra visibility in low light environments.
Certificaten: Conforms to BS 8442 and EN 12899-1.