Cough Guard 2 (fits bump caps)
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JSPAAHV970-001-100EA 26.2 EUR
JSPAAHV970-001-100
Cough Guard 2 (fits bump caps)
Gelaatsschermen
The Cough Guard can be fitted to a JSP bump cap to reduce the likelihood of exhaled bacteria/virus contacting the wearer's face. This device is not specifically certified and tested for COVID-19 protection.
  • Crystal clear APET 0,3 mm lens provides a layer of protection
  • The only JSP verified cough guard bump cap fitment
  • Quick and simple cleaning with soap and water
  • Prescription eyewear, face mask and ear defender compatible
Crystal clear APET 0,3 mm lens provides a layer of protection, helping to block aerosols and droplets. The device easily fits JSP Hardcap™ A1+ and Hardcap Aerolite and is suitable for all peak variants. The Cough Guard is also suitable for use without a bump cap. The 25 mm foam headband with an extra-wide ultrasonically welded elastic strap offers superior all day comfort.