Disposable moulded mask, Martcare®
Leverancier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BEG120-002-B00 BEG130-002-B00 BEH120-002-A00 BEH130-002-A00
JSPABEG120-002-B00EA 10.6 EUR
JSPABEG120-002-B00 JSPABEG130-002-B00 JSPABEH120-002-A00 JSPABEH130-002-A00
Disposable moulded mask, Martcare®
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
A range of polypropylene moulded disposable respirators, available in valved and unvalved options, offering a cost effective solution.
  • Inner foam nose bridge
  • Metal-free straps
  • Exhalation valve option
  • Cost effective
The Martcare® range of polypropylene moulded disposable respirators offers a cost-effective solution, subject to the rigorous batch testing and conformity procedures applicable to the entire JSP PPE range.
Mask straps are ultrasonically welded, ensuring no metal parts come into contact with the face. A foam area around the nose of the mask provides comfort and a firmer fit. A low resistance exhalation valve option is available for easy breathing.
FFP2 options offer 94% minimum filtration efficiency. FFP3 options offer 99% minimum filtration efficiency.
Certificaten: EN 149:2001+A1:2009.