Recycled cleaning cloths, r-MicronQuick
Leverancier: FHP VILEDA SCS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170605 170632 170633 170634
115-2962EA 19.2 EUR
115-2962 115-2963 115-2964 115-2965
Recycled cleaning cloths, r-MicronQuick
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
r-MicronQuick cleaning cloths help to further sustainability aims by reducing waste and CO2 emissions throughout the cleaning process. Highly durable r-MicronQuick cleaning cloths are made from up to 70% recycled superior micronfibre technology, for no compromises on the cleaning performance and hygiene.
  • Superior micronfibre cleaning performance
  • Made from 70% recycled PES
  • 99,99% virus removal performance
  • Streak-free cleaning
  • Can be used dry, damp, or wet
  • Cloths can withstand up to 400 wash cycles
r-MicronQuick cleaning cloths exhibit a very lowMore Product Information