Antistatic corrugated tubing, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R774-14 R775-13
BOHLR774-14EA 11.9 EUR
BOHLR774-14 BOHLR775-13
Antistatic corrugated tubing, b.safe
Slangen Chromatography Tubing
Flexible corrugated tubing.
  • Made from static dissipative plastics
Ordered quantities are supplied in continuous rolls by the metre.