Micro spatulas, USP Class VI, double-bagged, ULTRA
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9015H-170S 9015HN-170S 9015HV-170S
SAMP9015H-170SEA 193 EUR
SAMP9015H-170S SAMP9015HN-170S SAMP9015HV-170S
Micro spatulas, USP Class VI, double-bagged, ULTRA
Spatels
The double bagged micro spatula are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in class 100,000 cleanroom, packed in Class 10,000 cleanroom
Simply unwrap and use the micro spatula. The narrow design means they can be used with most vials and test tubes.
This range is ideal for sampling and dispensing small amounts.
Certificaten: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661.
Verpakking: Supplied in a lined box.
Order Now
Specificaties
Environmentally Preferable