Autoclavable bioreactor assemblies for microbial applications, BIOne
Leverancier: DISTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2022-1506 2022-1508 2022-1510
DISI2022-1506EA 0 EUR
DISI2022-1506 DISI2022-1508 DISI2022-1510
Autoclavable bioreactor assemblies for microbial applications, BIOne
Bioreactoren
The BIOne autoclavable bioreactor is available in a range of sizes including; 2, 5 and 10 L working volume. All metal components are manufactured from 316L stainless steel with electropolish.
Standard microbial configuration includes glass vessel, stainless steel head plate, vessel stand, dual-Rushton impellers, baffles, cooling finger, sparger, triple-addition port, overlay gas inlet port, exhaust condenser, harvest tube, thermowell, sample down tube, sensor ports, foam probe, septum port, addition ports and port plugs.
Bestelinformatie: If for use with a controller other than theMore Product Information