Disposable funnel for liquids, LaboPlast® / SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-6001 5378-6002 5379-0040 5379-1040 5378-6003 5378-6004
221-0303EA 17.5 EUR
221-0303 221-0304 221-0305 221-0306 221-0307 221-0314
Disposable funnel for liquids, LaboPlast® / SteriPlast®
Trechters Filtertrechters Filtertrechters
Disposable funnel for reliable filling of liquids, ideal for filling bottles and other containers with a narrow filler opening.
  • For easy and reliable filling or filtering
  • Suitable for use in laboratories and clean rooms or for sampling
  • Cleanroom manufactured (cleanroom Class 7)
  • Individually packaged and optionally sterilised
  • Complies with EU food requirements and FDA regulations
The disposable funnel ensures no loss when transferring liquid samples or free-flowing solids. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
Cleanroom produced funnel with suspension eye and venting fins for ventilation to ensure uniform flow while pouring. In combination with a filter paper the disposable funnel is also well suited for filtering.
The disposable funnel for liquids is ideal for use in laboratories and clean rooms or for sampling. It can be used in pharmaceuticals, chemistry and cosmetics and the food and animal feed industries, as well as all areas where special purity is important.
Verpakking: 10 pieces per pack, individually packaged.