Mini automated cell counter, Moxi™ Z
Leverancier: Orflo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORFLMXZ001EA 3720 EUR
ORFLMXZ001
Mini automated cell counter, Moxi™ Z
Celtellers Automatische celtellers
The Moxi™ Z automated cell counter combines the the 'gold-standard' Coulter Principle typically used in high-end instruments with a patented thin-film sensor technology to provide highly accurate and repeatable direct cell/particle size and count over a broad dynamic range (2 to 34 µm), with single cell resolution.
Through the utilization of state of the art semiconductor, embedded processing and software algorithms the Moxi™ Z yields results in 8 to 15 seconds, with little to no sample prep required. The Moxi™ Z also provides a proprietary and revolutionary new insight into your cell culture via the Moxi Population Index (MPI). Now, for the first time, cell culturists can instantly view the entire state of their culture and leverage this indicator as an in-lab QC tool to normalize and standardize their culture techniques. This is made possible by the power of Coulter and the Moxi™ Z proprietary algorithm, which provides a ratio of the primary population to all other particles, cell debris, or possible contamination in the sample.
The instrument allows for measurement of cells and particulates in a wide variety of applications, from mammalian cells, cancer cell lines, HUVEC’s, epithelial cells, endothelial cells, stem cells, mesenchymal stem cells (MSC’s), human embryonic stem cells (hESC’s), iPS (induced pluripotent stem cells), Red Blood Cells (RBC’s), White Blood Cells (WBC’s), Peripheral Blood Mononuclear Cells (PBMC’s) to common yeast, beads (both naked and died), and more.
The instrument is ultra-small and uses a long-life battery. It is ideal for use in the hood. And with the convenient Bluetooth connectivity feature, researchers can also use the powerful MoxiChart software package, or excel, to further analyze their data on a Mac or PC.
Due to it’s ease of use and the richness of the data output, the Moxi™ Z acts as a powerful front and back end tool to more complex highly sophisticated instrumentation such as flow cytometers, particle analyzers, and NGS (Next Gen Sequencers).
Moxi™ Z was designed to overcome the tedium associated with manual counting and the lack of repeatability of image-based counting systems.
Bestelinformatie: The kit includes the Moxi™ Z instrument, 25 cassettes (sufficient for 50 tests), Cassette Dispenser, Calibration Check Beads, USB cable/power adapter and USB flash drive containing User Guide and MoxiChart software. The cassettes are not reusable