Bowl, stainless steel, shallow form
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4208-0020 4208-0021 4208-0022 4208-0023 4208-0024 4208-0025
BURK4208-0020EA 17.8 EUR
BURK4208-0020 BURK4208-0021 BURK4208-0022 BURK4208-0023 BURK4208-0024 BURK4208-0025
Bowl, stainless steel, shallow form
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
The robust stainless steel bowl is ideal for cleaning, disinfecting or sterilising instruments as well as for placing and carrying different devices or samples. Due to the smooth surface, the instrument tray is easy to clean.
  • For cleaning, disinfecting, sterilising or placing away laboratory cutlery
  • Stable and hygienic
  • Autoclavable
The rectangular bowl has a low edge and can be used in the various areas of pharmacy, chemistry, cosmetics and food.