Xylene-Resistant Thermal Transfer Labels for Paraffin Wax Blocks
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRF-117C1-2WH PRF-117C3-4WH PRF-125C1-4WH PRF-125C3-4WH PRF-149C1-4WH PRF-149C3-10WH PRF-209C1-4WH PRF-209C3-4WH PRF-213C1-5WH PRF-213C3-10WH PRF242C1-0.5WH PRF-242C3-1WH PRF-509C1-4WH PRF-509C3-8WH PRF-67C1-1WH PRF-67C3-1WH PRF-7C1-1WH PRF-7C3-3WH
GAINPRF-117C1-2WHEA 139 EUR
GAINPRF-117C1-2WH GAINPRF-117C3-4WH GAINPRF-125C1-4WH GAINPRF-125C3-4WH GAINPRF-149C1-4WH GAINPRF-149C3-10WH GAINPRF-209C1-4WH GAINPRF-209C3-4WH GAINPRF-213C1-5WH GAINPRF-213C3-10WH GAINPRF242C1-0.5WH GAINPRF-242C3-1WH GAINPRF-509C1-4WH GAINPRF-509C3-8WH GAINPRF-67C1-1WH GAINPRF-67C3-1WH GAINPRF-7C1-1WH GAINPRF-7C3-3WH
Xylene-Resistant Thermal Transfer Labels for Paraffin Wax Blocks
Etiketten
These labels are intended for hard-to-stick surfaces, including paraffin wax, PTFE, Perfluoroalkoxy (PFA), and Parchment paper. These thermal transfer printable labels are chemical-resistant, perfect for labeling paraffin-embedded tissue cassettes used in histology applications.
  • Temperature range: −55 to +70 °C
  • Adheres to difficult surfaces, including paraffin wax blocks
  • Withstands exposure to harsh chemicals and solvents
  • Printable with all major barcode printers
  • XAR ribbon is recommended
Thermal transfer printable thermoplastic labels that adhere strongly to paraffin wax blocks and other hard-to-stick surfaces such as Polytetrafluoroethylene (PTFE), Perfluoroalkoxy (PFA), Parchment paper, etc. These labels are water- and chemical-resistant, ideal for identifying paraffin wax blocks for histology applications. A chemical-resistant printout is obtained when printed with the XAR-class of solvent-resistant ribbons. The labels and printed information will resist xylene, toluene, alcohol, and other solvents, as well as chemical exposure without additional protection.