Aluminium tool for vial transfer, CryoHolder
Leverancier: PHARSOL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PS-OO1-CH-T PS-OO1-CH-C
479-2294EA 0 EUR
479-2294 479-2293
Aluminium tool for vial transfer, CryoHolder
flesjes
The CryoHolder is a tool that enables you quick, more efficient and safe transfer of frozen cryovials. The tool is made out of aluminium so it can withstand big temperature changes; from –196 °C in liquid nitrogen up to 121 °C in the autoclave.
  • Compatible with all major cryovial brands
  • Pen sized and shaped
  • Very easy to use
The CryoHolder's ergonomic design allows faster workflow, eliminates both frostbite and damage to the biological material while making the transfer more efficient.
The CryoHolder is made out of aluminium, which enables working with liquid nitrogen as well as sterilising it in the autoclave at 121 °C. It is also resistant to mechanical damage that may occur during the work process.
The CryoHolder allows you to work faster, as you can easily pick up cryovials and move them around. Compared to using tweezers or forceps, the CryoHolder helps you complete the task 55% faster, thus making sure the biological sample doesn't thaw.
Model T is the model that fits vials and cryogenic tubes from the following manufacturers:
- Thermo Fisher Nunc vials
- Starlab vials
- CryoKING vials (external thread)
- SSIbio vials
- CELLTREAT Freeze vials (internal thread)
Model C is the model that fits vials and cryogenic tubes from the following manufacturers:
- Corning vials
- Elkay vials
- Simport vials
- Alpha Laboratories vials
- Greiner Bio-One vials
- VWR® vials
- Sarstedt CryoPure vials
- Daiger AvitroLab vials (internal thread)
Verpakking: Packaged as a single product in cardboard packaging.