Stand-up lectern tables, 304 stainless steel
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DELT-1SHMS DELT-1SHS DELT-2SHMS DELT-2SHS DELT-FL DELT-FX DELT-MS DELT-S
TEKMDELT-1SHMSEA 0 EUR
TEKMDELT-1SHMS TEKMDELT-1SHS TEKMDELT-2SHMS TEKMDELT-2SHS TEKMDELT-FL TEKMDELT-FX TEKMDELT-MS TEKMDELT-S
Stand-up lectern tables, 304 stainless steel
Meubilair Tafels
304 grade stainless steel lecterns with sloped top that are perfect for standing at. Durable and strong, with hygienic design features to minimise risk. They are fully-welded, with reduce dirt traps to reduce risk. Choose from freestanding, wall mounted or mobile, with a wide choice of shelf options too.
  • 304-grade EU sourced stainless steel
  • Supplied fully welded, no construction or assembly required
  • Ideal for standing
  • Inclined top with lip to hold documents in place
  • Hygienic design throughout
  • Brushed dull polish finish
  • Choice of mobile or static
  • Choice of no shelf, 1 shelf or 2 shelves
  • Minimised dirt traps
Sturdy 30 mm box section frame, designed with hygiene in mind, all dirt traps minimised from every angle.
Load capacity: 150 kg