Stand-up/sit down tables/workbench, height-adjustable
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ETSS1300X750 ETSS1300X750/E TLWB-AJMA16X75 ETSS1600X750/E ETSS1900X750 TLWB-AJEL19X75
TEKMETSS1300X750EA 0 EUR
TEKMETSS1300X750 TEKMETSS1300X750/E TEKMTLWB-AJMA16X75 TEKMETSS1600X750/E TEKMETSS1900X750 TEKMTLWB-AJEL19X75
Stand-up/sit down tables/workbench, height-adjustable
Meubilair Tafels
The height adjustable stainless steel table is manufactured from fully welded 304-grade stainless steel in a brushed dull polish finish and is available in electronic or manual wind height adjustment.
  • Free standing
  • 304-grade stainless steel
  • Brushed dull polish finish
  • Fully welded, no construction or assembly required
  • Hydraulic height adjustable table - manual or electric
  • Stainless steel feet with non-marking rubber base
  • 700 to 1000 mm height adjustment (table top height)
The 300 mm height adjustment, ensures a work surface comfortable for workers of different heights.
Load capacity: 250 kg