Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TSSC1200X600 TSSC1500X600 TSSC1500X750 TSSC1800X600 TSSC1800X750
TEKMTSSC1200X600EA 0 EUR
TEKMTSSC1200X600 TEKMTSSC1500X600 TEKMTSSC1500X750 TEKMTSSC1800X600 TEKMTSSC1800X750
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
Meubilair Tafels
Stainless steel computer desk wih pull out keyboard and support for PC tower, available in solid or perforated top for optimum air flow.
  • Computer desk with top and tower shelf - silicone seal
  • Fully welded 304 grade stainless steel, no construction or assembly required
  • Brushed dull polish finish
  • Solid or perforated top
  • Keyboard/mouse drawer
  • Stainless steel adjustable feet with non-marking rubber base
A ledge for a regular size computer tower and a keyboard shelf, also manufactured from 304 grade stainless steel, this desk is designed to house your computer safely.
Load capaciity: 250 kg