Self-laminating vinyl labels for M210, M210-LAB, and M211 label printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
152083 152084
BRDY152083EA 33.98 EUR
BRDY152083 BRDY152084
Self-laminating vinyl labels for M210, M210-LAB, and M211 label printers
Etiketten
Popular wire and cable labels. Conformable material is designed to adhere to highly curved surfaces, wires, and cables.
  • Permanent acrylic adhesive
  • Rounded corners
  • Suitable for freezer and liquid nitrogen
  • For smooth and textured/rough surfaces
  • Service temperature: −70 to 70 °C
  • For use with M210 and M211 portable label printers
Labels feature a white printable area with clear 'tail' that wraps around the marker. The clear tail serves as over-laminate to protect the text, providing superior resistance to abrasion, dirt, oils, solvents, water, alcohol, and excessive handling.