Sampling bags, SteriBag Cleanroom
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5344-8010 5344-8012
BURK5344-8010EA 426 EUR
BURK5344-8010 BURK5344-8012
Sampling bags, SteriBag Cleanroom
Zakken Monsterzakken
The sampling bag SteriBag Cleanroom with double packaging can be placed directly in the cleanroom. 25 sample bags are placed in double secondary packaging. They can be removed one after the other and are therefore ideally suited for introducing into the cleanroom.
  • Sterile, with double packaging
  • RNase, DNase and pyrogen-free
  • PE, transparent
  • Liquid-tight
  • With labelling field
Then the sterile bags can be used directly in the clean room. SteriBag Cleanroom is sterile inside and outside. This guarantees contamination-free storage and transport of solid,More Product Information
Order Now
Specificaties
Environmentally Preferable