Round bottles with snap closure
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0308-1000 0308-0500 0308-0100 0308-0250
BURK0308-1000EA 3.4 EUR
BURK0308-1000 BURK0308-0500 BURK0308-0100 BURK0308-0250
Round bottles with snap closure
Flessen Kunststof flessen
Handy round bottles with snap-action hinged lid, especially suited for cosmetic liquids, emulsions, creams, and oils. Sampling possible per portion.
  • HDPE transparent bottle, PP closure
  • Snap closure with hole Ø 3 mm
  • Cost-effective and shapely