Table/workbench with solid shelf, 304 stainless steel, mobile or static
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MTSSU0600X600S MTSSU0900X600S MTSSU1200X600S MTSSU1200X750S MTSSU1500X600S MTSSU1500X750S MTSSU1500X900S MTSSU1800X600S MTSSU1800X750S MTSSU1800X900S MTSSUB0600600S MTSSUB0900600S MTSSUB1200600S MTSSUB1200750S MTSSUB1500600S MTSSUB1500750S MTSSUB1500900S MTSSUB1800600S MTSSUB1800750S TLWB1SUPM18X90 TSSU0600X600 TSSU0900X600 TSSU1200X600 TSSU1200X750 TSSU1500X600 TSSU1500X750 TSSU1500X900 TSSU1800X600/4 TSSU1800X750/4 TSSU1800X900/4 TSSUB0600X600 TSSUB0900X600 TSSUB1200X600 TSSUB1200X750 TSSUB1500X600 TSSUB1500X750 TSSUB1500X900 TSSUB1800X6004 TSSUB1800X7504 TSSUB1800X9004
TEKMMTSSU0600X600SEA 0 EUR
TEKMMTSSU0600X600S TEKMMTSSU0900X600S TEKMMTSSU1200X600S TEKMMTSSU1200X750S TEKMMTSSU1500X600S TEKMMTSSU1500X750S TEKMMTSSU1500X900S TEKMMTSSU1800X600S TEKMMTSSU1800X750S TEKMMTSSU1800X900S TEKMMTSSUB0600600S TEKMMTSSUB0900600S TEKMMTSSUB1200600S TEKMMTSSUB1200750S TEKMMTSSUB1500600S TEKMMTSSUB1500750S TEKMMTSSUB1500900S TEKMMTSSUB1800600S TEKMMTSSUB1800750S TEKMTLWB1SUPM18X90 TEKMTSSU0600X600 TEKMTSSU0900X600 TEKMTSSU1200X600 TEKMTSSU1200X750 TEKMTSSU1500X600 TEKMTSSU1500X750 TEKMTSSU1500X900 TEKMTSSU1800X600/4 TEKMTSSU1800X750/4 TEKMTSSU1800X900/4 TEKMTSSUB0600X600 TEKMTSSUB0900X600 TEKMTSSUB1200X600 TEKMTSSUB1200X750 TEKMTSSUB1500X600 TEKMTSSUB1500X750 TEKMTSSUB1500X900 TEKMTSSUB1800X6004 TEKMTSSUB1800X7504 TEKMTSSUB1800X9004
Table/workbench with solid shelf, 304 stainless steel, mobile or static
Meubilair Tafels
This stainless steel workbench features a solid undershelf, perfect for storing smaller items out of the way, but close to hand for when they are needed. Durable and strong, this range is ideal for heavier payloads. Fully welded, they reduce the risk of dirt traps.
  • 304 grade stainless steel; brushed dull polish finish
  • Supplied fully welded, no assembly or construction required
  • Solid undershelf
  • Choose from static or mobile - BZP or stainless steel castors
  • Added strength
304 grade stainless steel, Sturdy 40 mm box section frame, designed with hygiene in mind.
Load capacity: 225 kg (for models with castors) and 250 kg (for models with feet).