Clip for tubes and cables, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1402-05 R1403-05
BOHLR1402-05EA 24.9 EUR
BOHLR1402-05 BOHLR1403-05
Clip for tubes and cables, b.safe
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
For easy bundling and fixing of signal cables, grounding cables, waste tubes, and vent tubes inside of safety cabinets and built-in installations.
  • Chemical resistance: ++ very good
  • Material: POM
  • Temperature resistance: from −30 to +100 °C
Clip made of mechanically tight POM for safe clipping of the different cables or tubes. Design for wall mount includes fastening material.