Ultra fine smear-proof markers
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PEN-61IR PEN-62IR
MICRPEN-61IREA 197 EUR
MICRPEN-61IR MICRPEN-62IR
Ultra fine smear-proof markers
Pennen Cleanroompennen
A permanent marker that gives the thinnest permanent line at 0,3 mm.
  • Long nib allows for writing in limited spaces
  • Works well with lab samples and slides, glass, plastic, and paper
  • Gamma irradiated for aseptic environments
  • Available in black or blue ink
Writes permanently on most surfaces with quick drying ink that is fade- and water-resistant to further extend multiple project uses. Designed for documentation and labeling in aseptic environments.