ESD safety shoes, lace-up, TERENCE XXG Low, S3 HI
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
728631/39 728631/40 728631/41 728631/42 728631/43 728631/44 728631/45 728631/46 728631/47 728631/48
ELTN728631/39EA 111.9 EUR
ELTN728631/39 ELTN728631/40 ELTN728631/41 ELTN728631/42 ELTN728631/43 ELTN728631/44 ELTN728631/45 ELTN728631/46 ELTN728631/47 ELTN728631/48
ESD safety shoes, lace-up, TERENCE XXG Low, S3 HI
Schoenen en laarzen
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole WELLMAXX Grip
  • Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.
Certificaten: EN ISO 20345 S3 HI/HRO/SRC, form A