ESD safety ankle boots, DIALUTION, Mid, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
760511/36 760511/37 760511/38 760511/39 760511/40 760511/41 760511/42 760511/43 760511/44 760511/45 760511/46 760511/47 760511/48
ELTN760511/36EA 91.9 EUR
ELTN760511/36 ELTN760511/37 ELTN760511/38 ELTN760511/39 ELTN760511/40 ELTN760511/41 ELTN760511/42 ELTN760511/43 ELTN760511/44 ELTN760511/45 ELTN760511/46 ELTN760511/47 ELTN760511/48
ESD safety ankle boots, DIALUTION, Mid, S3
Schoenen en laarzen
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO grey
  • Non-metallic, penetration-resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.