ESD safety ankle boots, BRAYDEN XXTM, Strap Closure, Mid, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
760531/36 760531/37 760531/38 760531/39 760531/40 760531/41 760531/42 760531/43 760531/44 760531/45 760531/46 760531/47 760531/48
ELTN760531/36EA 92.9 EUR
ELTN760531/36 ELTN760531/37 ELTN760531/38 ELTN760531/39 ELTN760531/40 ELTN760531/41 ELTN760531/42 ELTN760531/43 ELTN760531/44 ELTN760531/45 ELTN760531/46 ELTN760531/47 ELTN760531/48
ESD safety ankle boots, BRAYDEN XXTM, Strap Closure, Mid, S3
Schoenen en laarzen
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO grey
  • Non-metallic, penetration-resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Metal- and leather-free.