Safety ankle boots, RENEGADE Work GTX, Mid, S3 CI
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5652/39 5652/40 5652/41 5652/42 5652/43 5652/44 5652/45 5652/46 5652/47
114-9426EA 157.9 EUR
114-9426 114-9427 114-9428 114-9429 114-9430 114-9431 114-9432 114-9433 114-9434
Safety ankle boots, RENEGADE Work GTX, Mid, S3 CI
Schoenen en laarzen
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip in the shoe.
  • Hydrophobised nubuck leather/Hydrophobized textile material
  • GORE-TEX Performance Comfort Footwear
  • Full, padded bellows tongue
  • Full-length inlay sole LOWA
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole VIBRAM® with MONOWRAP®-technology
  • Steel toe cap
The tongue prevents pressure marks and dirt from entering into the shoe. The heel loop makes it easier to get inside the shoe.