ESD safety ankle boots, FINJA XXTL, Mid, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
746181/35 746181/36 746181/37 746181/38 746181/39 746181/40 746181/41 746181/42
ELTN746181/35EA 95.9 EUR
ELTN746181/35 ELTN746181/36 ELTN746181/37 ELTN746181/38 ELTN746181/39 ELTN746181/40 ELTN746181/41 ELTN746181/42
ESD safety ankle boots, FINJA XXTL, Mid, S3
Schoenen en laarzen
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO Lady
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Trainers Lady
  • Abrasion-proof toe protection
  • Steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.