ESD safety shoes, BRICE XXTM, low, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720551/36 720551/37 720551/38 720551/39 720551/40 720551/41 720551/42 720551/43 720551/44 720551/45 720551/46 720551/47 720551/48
ELTN720551/36EA 84.9 EUR
ELTN720551/36 ELTN720551/37 ELTN720551/38 ELTN720551/39 ELTN720551/40 ELTN720551/41 ELTN720551/42 ELTN720551/43 ELTN720551/44 ELTN720551/45 ELTN720551/46 ELTN720551/47 ELTN720551/48
ESD safety shoes, BRICE XXTM, low, S3
Schoenen en laarzen
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO grey
  • Non-metallic, penetration-resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Metal- and leather-free.