ESD safety shoes, MADDIE XXTL, low, S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
742331/35 742331/36 742331/37 742331/38 742331/39 742331/40 742331/41 742331/42
ELTN742331/35EA 91.9 EUR
ELTN742331/35 ELTN742331/36 ELTN742331/37 ELTN742331/38 ELTN742331/39 ELTN742331/40 ELTN742331/41 ELTN742331/42
ESD safety shoes, MADDIE XXTL, low, S3
Schoenen en laarzen
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre/Hydrophobised textile material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD PRO Lady
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Trainers Lady
  • Steel toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free, 100% vegan.