VWR®, Screw Neck Vials ND13
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
548-0509AEA 399 EUR
548-0509A 548-0523A 548-0529A 548-0528A 548-0519A 548-0524A 548-0521A 548-0526A 548-0522A 548-0527A 548-0051A 548-0052A 548-0510A 548-1395A 548-3252A 548-1454A 548-3314A
VWR®, Screw Neck Vials ND13
flesjes
ND13 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
ND13 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with inserts. Associated screw caps, mostly available with assembled septa, are engineered to prevent sample contamination while providing consistent, secure seals. The use of closed caps allows the samples to be stored for a long time (long-term storage applications).