VWR®, Screw Neck Vials ND15
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
548-0889AEA 605 EUR
548-0889A 548-0820A 548-0821A 548-0903A
VWR®, Screw Neck Vials ND15
flesjes
Screw Neck ND15 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
Screw Neck ND15 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom. Associated screw caps mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. The use of closed caps allows the samples to be stored for a long time (long-term storage applications).