VWR®, Snap Cap Vials
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
548-0141AEA 575 EUR
548-0141A 548-0649A 548-0554A 548-0625A 548-0621A 548-0620A 548-0623A 548-0622A 548-0624A 548-0650A 548-0651A 548-0144A 548-0652A 548-0142A 548-0555A 548-0143A 548-0663A
VWR®, Snap Cap Vials
flesjes
Snap cap vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They should only be used for storage of solids and powders.
  • No liners needed in the cap
  • Good handling with gloves
  • Quick and easy open/close
  • Easy filling
Snap cap vials constructed from type I borosilicate glass with low metal content. Vials are available in clear version with a flat bottom without a cap or as a kit with a PE cap. The usage of these vials allows the samples to be stored for a long time (long-term storage applications).