1M tris-hydrochloride buffers, Corning®
Leverancier: Corning
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
46-030-CM
46-031-CM
MDTC46-030-CMEA
355.33
EUR
MDTC46-030-CM
MDTC46-031-CM
1M tris-hydrochloride buffers, Corning®
Buffers
Biologische buffers
Tris-HCl buffer has many applications in molecular biology work, including use in nucleic acid purification.
This product can be diluted to obtain the desired concentration.