DMEM/Ham's F-12 50/50 mix products, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90-091-PB 90-090-PB
90-091-PBEA 50.42 EUR
90-091-PB 90-090-PB
DMEM/Ham's F-12 50/50 mix products, Corning®
Celcultuurmedia
This 1:1 mixture of Dulbecco's Modified Eagle Medium (DMEM) and Ham's F-12 media is ideal for culture of a variety of cell types under low serum conditions
  • Low endotoxin - <0,25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
DMEM is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME.  Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.