Corning® cell expansion bags
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
91-200-84 91-200-85 91-200-86 91-200-87
MDTC91-200-84EA 93.37 EUR
MDTC91-200-84 MDTC91-200-85 MDTC91-200-86 MDTC91-200-87
Corning® cell expansion bags
Zakken Mediazakken
Corning® Cell Expansion Bags are intended for the expansion and culture of non-adherent cells.
  • Made from a single-web polyolefin, gas permeable film
  • Port 1 - Two leads with female Luers and pinch clamp
  • Port 2 - Dockable fill line with sampling valve and pinch clamp
The integrated tubing allows for functionally closed system filling, feeding, and sampling.